Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER81 Polyclonal Antibody, Abbomax

Catalog No. 502204123
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4123 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4123 Supplier Abbomax Inc Supplier No. 602930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Mouse brain Cellular location: Nuclear, Cytoplasmic.

ER81 Polyclonal antibody specifically detects ER81 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ER81
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Etv1
Gene Accession No. P41164, P50549
Gene Alias DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of human ER81 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14009, 2115, 362733
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.