Learn More
Invitrogen™ ERAB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579401
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human 293T whole cell, human Hela whole cell, human MOLT4 whole cell. IHC: Human Mammary Cancer tissue. ICC/IF: U20S cell, Hela Cell, MCF-7 Cell, MCF-7 Cell, SMMC-7721 Cell. Flow: A549 cell, A431 cell.
This gene encodes 3-hydroxyacyl-CoA dehydrogenase type II, a member of the short-chain dehydrogenase/reductase superfamily. The gene product is a mitochondrial protein that catalyzes the oxidation of a wide variety of fatty acids, alcohols, and steroids. The protein has been implicated in the development of Alzheimer's disease, and mutations in the gene are the cause of 2-methyl-3-hydroxybutyryl-CoA dehydrogenase deficiency (MHBD). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
Specifications
ERAB | |
Polyclonal | |
Unconjugated | |
HSD17B10 | |
17-beta-HSD 10; 17-beta-hydroxysteroid dehydrogenase 10; 17bHSD10; 17b-HSD10; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; ABAD; AB-binding alcohol dehydrogenase; Ads9; amyloid beta-peptide binding protein; amyloid-beta peptide binding alcohol dehydrogenase; CAMR; DUPXp11.22; endoplasmic reticulum-associated amyloid beta-peptide-binding protein; ERAB; HADH2; HADH2 protein; HCD2; HSD17B10; hydroxyacyl-Coenzyme A dehydrogenase type II; hydroxyacyl-Coenzyme A dehydrogenase, type II; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid 17-beta dehydrogenase 10; hydroxysteroid dehydrogenase 10; MHBD; mitochondrial ribo; Mitochondrial ribonuclease P protein 2; mitochondrial RNas; mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; MRPP2; MRX17; MRX31; MRXS10; RP3-339A18.2; SCHAD; SDR5C1; Short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; type 10 17b-HSD; Type II HADH; XH98G2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3028 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q99714 | |
HSD17B10 | |
E.coli-derived human ERAB recombinant protein (Position: E48-P261). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.