Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERAF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30322220UL
Description
ERAF Polyclonal specifically detects ERAF in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ERAF | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
51327 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
alpha hemoglobin stabilizing protein, EDRFalpha hemoglobin stabilising protein, ERAFalpha-hemoglobin-stabilizing protein, erythroid associated factor, erythroid differentiation associated factor, Erythroid differentiation-related factor, Erythroid-associated factor | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-102 of human ERAF (NP_057717.1). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction