Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555073 Supplier Invitrogen™ Supplier No. PA555073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVLVMNKVDC LKQKSVLLEL TAALTEGVVN GKKLKMRQAF HSHPGTHCPS PAVKDPNTQS VGNPQRIGWP HFKEIFMLSA LSQEDVKTLK QY Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.

The protein encoded by this gene is a GTPase that localizes to the mitochondrion. The encoded protein binds to the 3' terminal stem loop of 12S mitochondrial rRNA and is required for proper assembly of the 28S small mitochondrial ribosomal subunit. Deletion of this gene has been shown to cause mitochondrial dysfunction, growth retardation, and apoptosis. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERAL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERAL1
Gene Accession No. O75616
Gene Alias 2610524P08Rik; 9130407C09Rik; AU019798; CEGA; Conserved ERA-like GTPase; ERA; era (E. coli G-protein homolog)-like 1; Era (G-protein)-like 1; Era (G-protein)-like 1 (E. coli); Era G-protein-like 1; Era like 12S mitochondrial rRNA chaperone 1; Eral1; ERAL1A; Era-like 1; Era-like 12S mitochondrial rRNA chaperone 1; ERA-like GTPase; ERA-like protein 1; ERA-W; GTPase Era, mitochondrial; GTPase, human homolog of E. coli essential cell cycle protein Era; GTP-binding protein era homolog; HERA; H-ERA; HERA-A; HERA-B; Mera; M-ERA; MERA-S; MERA-W
Gene Symbols ERAL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERAL1. Recombinant protein control fragment (Product #RP-93314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26284
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.