Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAL1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569514
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9514 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9514 Supplier Bethyl Laboratories, Inc Supplier No. A305092AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The function remains unknown.

Specifications

Antigen ERAL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ERAL1
Gene Accession No. O75616
Gene Alias CEGA, conserved ERA-like GTPase, ERA, Era (E. coli G-protein homolog)-like 1, Era G-protein-like 1, Era G-protein-like 1 (E. coli), ERAL1A, Era-like 12S mitochondrial rRNA chaperone 1, ERA-like protein 1, ERA-W, GTPase, human homolog of E. coli essential cell cycle protein Era, GTP-binding protein era homolog, hERA, H-ERA, HERA-A, HERA-B
Gene Symbols ERAL1
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26284
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.