Learn More
ERAL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305092A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function remains unknown.Specifications
ERAL1 | |
Polyclonal | |
Unconjugated | |
ERAL1 | |
CEGA, conserved ERA-like GTPase, ERA, Era (E. coli G-protein homolog)-like 1, Era G-protein-like 1, Era G-protein-like 1 (E. coli), ERAL1A, Era-like 12S mitochondrial rRNA chaperone 1, ERA-like protein 1, ERA-W, GTPase, human homolog of E. coli essential cell cycle protein Era, GTP-binding protein era homolog, hERA, H-ERA, HERA-A, HERA-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26284 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O75616 | |
ERAL1 | |
Between 75 and 125 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.