Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAP2 Mouse anti-Human, Unconjugated, Clone: 2B11A2, Proteintech
SDP

Catalog No. 502239567 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9567 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9567 Supplier Proteintech Group Inc Supplier No. 674771IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The ERAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ERAP2. This antibody recognizes human, rat antigen. The ERAP2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen ERAP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B11A2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 64167
Target Species Human, Rat
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.