Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERAS Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299426JF646
Description
ERAS Polyclonal antibody specifically detects ERAS in Mouse samples. It is validated for Western BlotSpecifications
ERAS | |
Polyclonal | |
Western Blot | |
Embryonic stem cell-expressed Ras, E-Ras, ES cell expressed Ras, HRAS2MGC126693, HRASPGTPase ERas, MGC126691, small GTPase protein E-Ras, v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2, v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ERAS | |
0.1 mL | |
Signal Transduction | |
3266 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction