Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERAS Monoclonal Antibody (D10-B9)
GREENER_CHOICE

Catalog No. PIMA532907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532907 Supplier Invitrogen™ Supplier No. MA532907
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERAS Monoclonal Antibody for ICC/IF, IHC (P)

The human ERAS gene was initially reported as a processed pseudogene (HRasp, Ha-Ras2), with several base substitutions in coding regions, but was more recently described as potentially encoding a functional human ERAS protein. The ERAS oncogene supports the tumorigenic growth of embryonic stem cells by producing a constitutively active Ras protein. ERAS protein has less than 50% amino acid identity with H-Ras, another member of Ras family but the key domains essential for Ras function are highly conserved. The protein also contains amino-acid residues identical to those present in active mutants of Ras and causes oncogenic transformation in NIH 3T3 cells. Human ERAS gene is localized in the chromosomal region Xp11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERAS
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone D10-B9
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene ERAS
Gene Accession No. Q7Z444
Gene Alias ecat5; Embryonic stem cell-expressed Ras; ERAS; E-Ras; ES cell expressed Ras; ES cell-expressed Ras; GTPase ERas; Ha-Ras2; HRAS2; HRasp; small GTPase E-Ras; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene
Gene Symbols ERAS
Host Species Mouse
Immunogen Recombinant protein within Human ERAS aa 1-233.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.