Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAS Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
SDP

Catalog No. 502540757
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0757 30 μL
50-167-4528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0757 Supplier OriGene Supplier No. TA503689S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERAS Monoclonal antibody specifically detects ERAS in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen ERAS
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cnn2
Gene Accession No. Q99439
Gene Alias ecat5; Embryonic stem cell-expressed Ras; ERAS; E-Ras; ES cell expressed Ras; ES cell-expressed Ras; GTPase ERas; Ha-Ras2; HRAS2; HRasp; small GTPase E-Ras; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene
Host Species Mouse
Immunogen Full length human recombinant protein of human CNN2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1265
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.