Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERBB2, anti-Human, Clone: 4D5-8 (trastuzumab), Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00109
Specifications
ERBB2 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
CD340/HER-2/HER-2/neu/HER2/NEU/NGL/TKR1 | |
Original antibody is raised against A431 cells expressing EGFR. | |
RUO | |
2064 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Flow Cytometry | |
4D5-8 (trastuzumab) | |
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
ERBB2 | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction