Learn More
Invitrogen™ ErbB2 (HER-2) Monoclonal Antibody (2G11), eBioscience™

Description
The antibody (2G11) can be used to stain cryostat sections or cell smears. BMS120 is also suitable as primary antibody in staining for FACS analysis, BMS120FI can be used for direct staining of cells, BMS120BT can be used in a 2-step procedure using the biotin-(strept)avidin system. The epitope is located in the extracellular domain, no cross reactivity with EGF-R has been detected by immunoprecipitation. Both HER-2 oncogene amplification and oncoprotein overexpression have been correlated with breast, ovarian, gastric, lung, and other cancers. In 15-40 % of primary breast cancers, amplification of the HER-2 oncogen is found which is highly correlated with overexpression of the encoded protein and seems to play a major role especially during the initiation of ductal carcinomas. HER-2 overexpression is described as independent prognostic factor with greater predictive power than most of the currently used prognostic tools - especially in axillary lymph-node-positive breast cancer patients. 32 % of ovarian carcinomas overexpress the HER-2 oncoprotein. Survival of those patients is significantly worse compared with cases of normal HER-2 protein expression. Additionally, patients whose tumors have high HER-2 protein expression are significantly less likely to have a complete response to primary therapy. A correlation between HER-2 expression, and clinical outcome has been also demonstrated for head and neck, salivary gland and placental carcinomas. HER-2 is useful...
Specifications
Specifications
| Antigen | ErbB2 (HER-2) |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2G11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2-7.4 |
| Gene | ERBB2 |
| Gene Accession No. | P04626 |
| Gene Alias | avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.