Learn More
Invitrogen™ ErbB2 (HER-2) Recombinant Rabbit Monoclonal Antibody (SP3)
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.
Specifications
Specifications
| Antigen | ErbB2 (HER-2) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Clone | SP3 |
| Concentration | 0.122 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.2-7.4 |
| Gene | ERBB2 |
| Gene Accession No. | P04626 |
| Gene Alias | avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.