Learn More
Invitrogen™ ErbB2 (HER-2) Recombinant Rabbit Monoclonal Antibody (SP3)
Rabbit Recombinant Monoclonal Antibody
$1039.00 - $1573.00
Specifications
Antigen | ErbB2 (HER-2) |
---|---|
Clone | SP3 |
Concentration | 0.122 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MA139544
|
Invitrogen™
MA139544 |
1 mL |
Each for $1,573.00
|
|
|||||
PIMA516348
|
Invitrogen™
MA516348 |
500 μL |
Each for $1,039.00
|
|
|||||
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.Specifications
ErbB2 (HER-2) | |
0.122 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P04626 | |
2064 | |
Recombinant protein encoding extracellular domain of human c-erbB2. | |
Antibody |
SP3 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
ERBB2 | |
Primary | |
Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.