Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB2/Her2 Antibody (4D5-8 (trastuzumab)), DyLight 680, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275895FR
Description
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.Specifications
ErbB2/Her2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ERBB2 | |
A431 cells (human epidermoid carcinoma) (over)expressing EGFR. | |
0.1 mL | |
Primary | |
Oncogenic protein Her-2/neu (erbB-2), a member of the family of epidermal growth factor (EGF) receptors, overexpressed in 30% of invasive breast cancers and 70% of ductal carcinomata in situ, in a small percentage of melanomas, and in other malignancies originating from various organs, including ovary, kidney, colon, and bladder. | |
Store at 4°C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry | |
4D5-8 (trastuzumab) | |
Flow Cytometry, ELISA, Immunohistochemistry | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Human | |
Protein A purified | |
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
2064.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction