Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB2/Her2 Antibody (ERB2/776), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2449830.02MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| ErbB2/Her2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| P04626, P04626 | |
| ERBB2 | |
| Recombinant intracellular domain of human HER-2 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This MAb is specific to c-erbB-2/HER-2 and shows minimal cross-reaction with other members of the family. C-erbB-2/HER-2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary. | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| ERB2/776 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| Mouse | |
| 185 kDa | |
| 0.02 mg | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| 2064 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction