Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB2/Her2 Antibody (HRB2/282), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234642MFV450
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Flow CytometrySpecifications
ErbB2/Her2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
HRB2/282 | |
Flow Cytometry | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Recombinant extracellular domain of human ErbB2/Her2 protein (Uniprot: P04626) | |
0.1 mL | |
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
2064 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction