Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB2/Her2 Rabbit anti-Human, Clone: 511, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | ErbB2/Her2 |
|---|---|
| Clone | 511 |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Sandwich ELISA Capture 1:250-1:2000 |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
Description
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Capture.Specifications
| ErbB2/Her2 | |
| Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| Recombinant Human ErbB2/Her2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 511 | |
| Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| 0.2 um filtered solution in PBS | |
| 2064 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title