Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERBB2 Mouse anti-Human, Biotin, Clone: OTI1A12, liquid, TrueMAB™
SDP

Catalog No. 501677756
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7756 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7756 Supplier OriGene Supplier No. TA700241
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ErbB2 (Her2/neu) is a 185 kDa transmembrane receptor tyrosine kinase from the EGFR (epidermal growth factor receptor) family that regulates biological responses such as cell growth, differentiation, and tissue development. ErbB2 has no ligand binding domain of its own and cannot bind growth factors. However, ErbB2 does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signaling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Autophosphorylation of tyrosine 1139 on ErbB2 allows binding of Grb2 and the Src SH2 domain, which allows activation of the Ras, Raf, ERK1/2 and JAK/STAT signaling pathways, respectively. Alternative splicing of ErbB2 results in several transcript variants, some encoding different isoforms and others that have not been fully characterized. In humans, the gene encoding ERBB2 is present on chromosome 17. Amplification and/or overexpression of the ErbB2 gene has been reported in ovarian tumors, breast, lung and skin cancer.

Specifications

Antigen ERBB2
Classification Monoclonal
Clone OTI1A12
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene ERBB2
Gene Accession No. P04626
Gene Alias CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1
Gene Symbols ERBB2
Host Species Mouse
Immunogen Full length human recombit protein of human ERBB2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.