Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERBB2 Mouse anti-Human, Unconjugated, Clone: ERBB2/2452, Abnova™

Catalog No. 89337424 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-424 Supplier Abnova Corporation Supplier No. MAB20929
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ERBB2 Monoclonal antibody specifically detects ERBB2 in Human samples. It is validated for IHC-P.

Specifications

Antigen ERBB2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ERBB2/2452
Conjugate Unconjugated
Description v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Gene ERBB2
Gene Alias CD340, HER-2, HER-2/neu, HER2, NEU, NGL, TKR1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 311-462 of human ERBB2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.