Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ErbB3 Chimeric Recombinant Human Monoclonal Antibody (mh908)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA543072
This item is not returnable.
View return policy
Description
Specificity: Human HER3/ErbB3. This product is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The antibody was produced using recombinant antibody technology.
ErbB3 (c-erbB-3/HER-3) is a member of the type I family of growth factor receptors and binds to ligands in the heregulin family. ErbB3 is over-expressed in a variety of tumors in the prostate, bladder, breast, stomach, pancreas, and colon. Heregulin and EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents. The ErbB3 gene is located on the long arm of human chromosome 12 (12q13) and is transcribed into a 6. 2kb mRNA which is translated into a 160/180 kDa glycoprotein. The ErbB3 gene encodes a transmembrane receptor tyrosine kinase due to alternative splicing and forms heterodimers with other EGF receptor family members that have kinase activity. Heterodimerization leads to the activation of pathways that lead to cell proliferation or differentiation. Alternate transcriptional splice variants encoding different isoforms of ErbB3 have been characterized. One ErbB3 isoform lacks the intermembrane region, is secreted outside the cell, and acts to modulate the activity of the membrane-bound form. Additional splice variants have also been reported, but they have not been thoroughly characterized.
Specifications
ErbB3 Chimeric | |
Recombinant Monoclonal | |
1.62 mg/mL | |
PBS with no preservative | |
P21860 | |
ERBB3 | |
Recombinant Human HER3/ErbB3 protein. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Neutralization | |
mh908 | |
Unconjugated | |
ERBB3 | |
avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
Human | |
Protein A | |
RUO | |
2065 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction