Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB3/Her3 Antibody (OTI3E10), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270661R
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
| ErbB3/Her3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ERBB3 | |
| Full length recombinant protein of human ErbB3/Her3 (NP_001973) produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Immunohistochemistry | |
| OTI3E10 | |
| Immunohistochemistry | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2065.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction