Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB3/Her3 Rabbit anti-Human, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289334100UL
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for ELISA.Specifications
ErbB3/Her3 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
16 | |
ELISA 1:5000-1:10000 | |
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
Recombinant Human ErbB3/Her3 Protein | |
100 μg | |
Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
2065 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction