Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB3/Her3 Rabbit anti-Human, Clone: 5Z1F0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
Antigen | ErbB3/Her3 |
---|---|
Clone | 5Z1F0 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Western BlotSpecifications
ErbB3/Her3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2065 | |
IgG | |
Affinity purified |
5Z1F0 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
A synthetic phosphorylated peptide around Y1222 of human ErbB3/Her3 (P21860). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title