Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB3 Monoclonal Antibody (H3.105.5 (Ab105)), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA513008
Description
MA5-13008 targets HER-3 in WB, flow cytometry, ICC/IF, and IP applications and shows reactivity with Human and mouse samples. The MA5-13008 immunogen is extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein.
ErbB3 Monoclonal antibody specifically detects ErbB3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
ErbB3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P21860, Q61526 | |
ERBB3 | |
Extracellular domain of recombinant human c-erbB-3/HER-3 oncoprotein | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
H3.105.5 (Ab105) | |
Unconjugated | |
ERBB3 | |
avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
Mouse | |
Protein G | |
RUO | |
13867, 2065 | |
4° C | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction