Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ErbB4/Her4 Antibody (182818) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11311W
Description
ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
ErbB4/Her4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
182818 | |
Flow Cytometry, CyTOF-ready | |
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4, Glu26-Arg649, Accession # Q15303 | |
0.1 mL | |
Angiogenesis, Cancer, Protein Kinase, Tumor Suppressors | |
2066 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction