Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4 Monoclonal Antibody (H4.72.8 (Ab72)), Invitrogen™

Catalog No. PIMA513016
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513016 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513016 Supplier Thermo Scientific Supplier No. MA513016

Mouse Monoclonal Antibody

MA5-13016 targets HER-4 in WB, FACS applications and shows reactivity with Human and Mouse samples. The MA5-13016 immunogen is extracellular domain of recombinant human c-erbB-4/HER-4 oncoprotein.

Oncogene ERBB4 (HER4) is a member of the type I receptor protein tyrosine subfamily that includes EGFR, ERBB2, and ERBB3. ERBB4 binds and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. ErbB 4 is activated by neuregulins and NTAK (neural and thymus derived activator for ErbB kinases). ErbB 4 is a receptor for heregulin and is capable of mediating HGL-stimulated tyrosine phosphorylation of itself. Like other protein kinases, ErbB 4 mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. ErbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3. ErbB 4 levels have been found to be elevated in certain human tumor cell lines suggesting that it may play a role in some malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ErbB4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone H4.72.8 (Ab72)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene ERBB4
Gene Accession No. Q15303, Q61527
Gene Alias 4ICD; ALS19; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4; avian erythroblastosis oncogene B 4; c-erbB-4; erb-b2 receptor tyrosine kinase 4; Erbb4; ERBB4 intracellular domain; ERBB4 transcript variant I12DEL; ERBB4 transcript variant I20DEL; Her4; human epidermal growth factor receptor 4; Mer4; OTTHUMP00000209432; p180erbB4; proto-oncogene-like protein c-ErbB-4; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-4; s80HER4; Tyro-2; Tyrosine kinase-type cell surface receptor HER4; v-erb-a erythroblastic leukemia viral oncogene homolog 4; v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian); v-erb-b2 avian erythroblastic leukemia viral oncogene 4; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4
Gene Symbols ERBB4
Host Species Mouse
Immunogen Extracellular domain of recombinant human c-erbB-4/HER-4 oncoprotein
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13869, 2066
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.