Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERBB4 Rabbit anti-Human, Unconjugated, Clone: HC-5, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20016
Description
ERBB4 Monoclonal antibody specifically detects ERBB4 in Human samples. It is validated for WB, IP, ICC, Flow Cyt.Specifications
| ERBB4 | |
| Monoclonal | |
| Unconjugated | |
| ERBB4 | |
| Rabbit | |
| 100 μg | |
| 2066 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
| HC-5 | |
| v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| HER4, MGC138404, p180erbB4 | |
| A synthetic peptide corresponding to human ERBB4 | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction