Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Erbin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566288 Supplier Invitrogen™ Supplier No. PA566288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SENLKHIVNHD DVFEESEELS SDEEMKMAEM RPPLIETSIN QPKVVALSNN KKDDTKETDS LSDEVTHNSN QNNSNCSSPS Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erbin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERBIN
Gene Accession No. Q96RT1
Gene Alias 1700028E05Rik; densin-180-like protein; epididymis secretory protein Li 78; Erbb2 interacting protein; ERBB2 interacting protein-like; erbb2-interacting protein; Erbb2ip; Erbin; HEL-S-78; Kiaa1225; Lap2; mKIAA1225; Protein LAP2; RGD1562952; similar to Erbb2 interacting protein isoform 2
Gene Symbols ERBIN
Host Species Rabbit
Immunogen Recombinant Human Erbin. Recombinant protein control fragment (Product #RP-105141).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.