Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erbium(III) chloride, anhydrous, 99.9% (metals basis)
Click to view available options
Quantity:
10 g
50 g
Description
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1500°C |
| Melting Point | 774°C |
| Physical Form | Beads |
| Quantity | 50 g |
| Solubility Information | Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF. |
| Formula Weight | 273.62 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 233-385-0 |
| RTECSNumber | KD8575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction