Erbium(III) oxide, 99.9% (REO), Alfa Aesar™, REacton™

Manufacturer: Alfa Aesar 1131022

Quantity 100g

 View more versions of this product

Catalog No. AA1131022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12061-16-4
    Physical Form Powder
    Assay Percent Range 99.9% (REO)
    Molecular Formula Er2O3
    Formula Weight 382.56
    MDL Number MFCD00010991
    Chemical Name or Material Erbium(III) oxide

    Specifications

    Formula Weight 382.56
    Quantity 100g
    EINECS Number 235-045-7
    Physical Form Powder
    Density 8.64g/mL
    Assay Percent Range 99.9% (REO)
    Molecular Formula Er2O3
    MDL Number MFCD00010991
    CAS 12061-16-4
    Chemical Name or Material Erbium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit