Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade
Click to view available options
Quantity:
∼2 g
∼50 g
10 g
Description
Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Pink |
| Physical Form | Liquid |
| Quantity | ∼50 g |
| Solubility Information | Micible with water. |
| Formula Weight | 573.70 (465.61 Anhydrous) |
| Grade | Reagent |
| Odor | Odorless |
| Appearance | Pink liquid |
| Chemical Name or Material | Erbium(III) perchlorate hexahydrate |
Safety and Handling
| Hazard Category | H271-H315-H319-H335 |
| Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1, Packing Group: II |
| EINECSNumber | 237-840-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction