Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium powder, -40 mesh, 99.9% (REO)

Catalog No. AA4416914
Click to view available options
Quantity:
5 g
25 g

CAS: 7440-52-0 | Er | 167.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Color Gray
Quantity 25 g
Mesh Size −40 Mesh
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3089
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Erbium powder
Hazard Category H228-H260
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-160-1
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.