Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597221 Supplier Invitrogen™ Supplier No. PA597221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIALEQKKEE CLKMESQLKK AHEAALEARA SPEMSDRIQH LEREITRYKD ESSKAQAEVD RLLEILKEVE NEKNDKDKKI AELERQVKDQ NKKVANLKHK EQVEKKKSAQ MLEEARRRED NLNDSSQQLQ DSLRKKDDRI EELEEALRES V; Positive Samples: HepG2, Jurkat, NIH/3T3, Rat brain; Cellular Location: Cytoplasm, Golgi apparatus membrane, Membrane, Peripheral membrane protein.

The protein encoded by this gene is a member of a family of RIM-binding proteins. RIMs are active zone proteins that regulate neurotransmitter release. This gene has been found fused to the receptor-type tyrosine kinase gene RET by gene rearrangement due to the translocation t(10;12)(q11;p13). Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ERC1
Gene Accession No. Q811U3, Q8IUD2, Q99MI1
Gene Alias 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST; CAST2; CAZ-associated structural protein 2; Elks; ELKS/Rab6-interacting/CAST family member 1; ERC; ERC protein 1; Erc1; ERC-1; ERC1b; FLJ31750; KIAA1081; MGC12974; mKIAA1081; OTTHUMP00000160633; Rab6 interacting protein 2; RAB6-interacting protein 2; Rab6ip2; RAB6IP2A; RAB6IP2B; RIM-Binding Protein
Gene Symbols ERC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-850 of human ERC1 (NP_8298841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111173, 23085, 266806
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.