Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Erc2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584939
Description
Immunogen sequence: TQRDLNHLLQ QESGNRGAEH FTIELTEENF RR.
ERC2 is a protein coding gene.
Specifications
Erc2 | |
Polyclonal | |
Unconjugated | |
Erc2 | |
6430531D06; CAST; CAST1; CAST1/ERC2; CAZ-associated structural protein; CAZ-associated structural protein 1; Cmbp; cytomatrix protein p110; D14Ertd171e; ELKS/RAB6-interacting/CAST family member 2; ELKS2alpha; ELKSL; ERC protein 2; ERC2; KIAA0378; SPBC110; Spc110 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15083 | |
Erc2 | |
Recombinant protein corresponding to Human ERC2. Recombinant protein control fragment (Product #RP-108264). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction