Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Erc2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590725 Supplier Invitrogen™ Supplier No. PA590725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HNQQLEKKKN AQLLEEVRRR EDSMADNSQH LQIEELMNAL EKTRQELDAT KARLASTQQS LAEKEAHLAN LRIERRKQLE E; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell junction, Cytoplasm, cytoskeleton, synapse, synaptosome.

ERC2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erc2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Erc2
Gene Accession No. Q6PH08, Q8K3M6
Gene Alias 6430531D06; CAST; CAST1; CAST1/ERC2; CAZ-associated structural protein; CAZ-associated structural protein 1; Cmbp; cytomatrix protein p110; D14Ertd171e; ELKS/RAB6-interacting/CAST family member 2; ELKS2alpha; ELKSL; ERC protein 2; ERC2; KIAA0378; SPBC110; Spc110
Gene Symbols Erc2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 770-850 of human ERC2 (NP_0563911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238988, 259269
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.