Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERC2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30515620UL
Description
ERC2 Polyclonal specifically detects ERC2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ERC2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
26059 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
CAST, CAST1, CAZ-Associated Structural Protein, Cytomatrix Protein P110, ELKS/RAB6-Interacting/CAST Family Member 2, ELKSL, ERC Protein 2, KIAA0378, SPBC110, Spc110 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 770-850 of human ERC2 (NP_056391.1). | |
20 μL | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction