Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Antibody (8F1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 15 publications
Supplier: Novus Biologicals NB500704SS
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
ERCC1 Monoclonal specifically detects ERCC1 in Human, Rat, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ERCC1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200 | |
| P07992 | |
| ERCC1 | |
| Full length recombinant human ERCC1. [UniProt# P07992] | |
| 0.025 mL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2067 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8F1 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat, Chinese Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction