Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Antibody (ERCC1/2683), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308904AF647
Description
ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein ArraySpecifications
ERCC1 | |
Monoclonal | |
Alexa Fluor 647 | |
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary) (Uniprot: P07992) | |
100 μg | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2067 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Protein Array | |
ERCC1/2683 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction