Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Antibody (ERCC1 3H11), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100117AF750
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ERCC1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ERCC1 | |
Full length HIS-tagged recombinant human ERCC1 | |
0.1 mL | |
2067 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
ERCC1 3H11 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction