Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Antibody (OTI2H1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270685H
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ERCC1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ERCC1 | |
Full length recombinant protein of human ERCC1 (NP_001974) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2H1 | |
Western Blot, Immunohistochemistry | |
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
Mouse | |
Affinity Purified | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2067.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction