Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Antibody (OTI2H1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245501
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ERCC1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20 | |
Mouse | |
32.4 kDa | |
0.1 mL | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
2067 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2H1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
ERCC1 | |
Full length human recombinant protein of human ERCC1 (NP_001974) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction