Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Monoclonal Antibody (OTI12F11), TrueMAB™, OriGene
SDP

Catalog No. 502540381
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0381 30 μL
50-167-5051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0381 Supplier OriGene Supplier No. TA504333S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ERCC1 Monoclonal antibody specifically detects ERCC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen ERCC1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI12F11
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DTNB
Gene Accession No. O60941, O70585, P84060
Gene Alias COFS4; DNA excision repair protein ERCC-1; ERCC excision repair 1, endonuclease non-catalytic subunit; Ercc1; Ercc-1; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); hypothetical protein LOC405807; RAD10; UV20; zgc:77511
Host Species Mouse
Immunogen Full length human recombinant protein of human DTNB produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13528, 1838, 362715
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.