Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Monoclonal Antibody (OTI2A7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA501182

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-2324


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3′ of the DNA lesion while the ERCC1-XPF complex makes the 5′ incision.
Specifications

Specifications

ERCC1
Monoclonal
0.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P07992
ERCC1
Full length human recombit protein of human ERCC1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2A7
Unconjugated
ERCC1
COFS4, RAD10, UV20
Mouse
Affinity Chromatography
RUO
2067
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.