Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Monoclonal Antibody (OTI6H7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805691

Description
The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3′ of the DNA lesion while the ERCC1-XPF complex makes the 5′ incision.Specifications
ERCC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P07992 | |
ERCC1 | |
Full length human recombit protein of human ERCC1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6H7 | |
Unconjugated | |
ERCC1 | |
COFS4, RAD10, UV20 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2067 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction