Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Mouse anti-Human, Clone:1E5B3, Abnova™

Catalog No. 89333145 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-145 Supplier Abnova Corporation Supplier No. MAB16695
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human ERCC1.

Specifications

Antigen ERCC1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1E5B3
Conjugate Unconjugated
Description excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ERCC1
Gene Alias COFS4, UV20
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 151-297 of human ERCC1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2067
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.