Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984079
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4079 Supplier Bioss Supplier No. BS1726R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Human ERCC1 is an IgG2a Mouse Monoclonal Antibody that recognizes Excision Cross Complementing 1 (ERCC1), a human endonuclease. ERCC1 protein forms a complex with Xeroderma Pigmentosum Group F (XPF) protein. During nucleotide excision repair of damaged DNA, the ERCC1-XPF complex forms an incision at the 5’ end of the DNA lesion. Deficiencies in the nucleotide excision repair pathway result in disorders such as Xeroderma pigmentosum, Cockayne’s syndrome and trichothiodystrophy.

Specifications

Antigen ERCC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ERCC1
Gene Accession No. P07992
Gene Alias BOS_17715; COFS4; DNA excision repair protein ERCC-1; ERCC excision repair 1, endonuclease non-catalytic subunit; Ercc1; Ercc-1; excision repair cross-complementation group 1; excision repair cross-complementing 1; excision repair cross-complementing 1 isofrom 2-like; excision repair cross-complementing rodent repair deficiency, complementation group 1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); hypothetical protein LOC405807; LOC100348713; RAD10; UV20; zgc:77511
Gene Symbols ERCC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ERCC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2067
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.