Learn More
ERCC3 Mouse anti-Human, Clone: OTI6H8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809371

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.Specifications
ERCC3 | |
Monoclonal | |
Unconjugated | |
ERCC3 | |
BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB | |
Mouse | |
Affinity Chromatography | |
RUO | |
2071 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6H8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P19447 | |
ERCC3 | |
Human recombit protein fragment corresponding to amino acids 191-436 of human ERCC3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.