Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105801AP150UL

Description
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.Specifications
ERCC3 | |
Polyclonal | |
Unconjugated | |
ERCC3 | |
BTF2, BTF2 p89, ERCC3, GTF2H, RAD25, TFIIH, TFIIH 89 kDa subunit, TFIIH p89, XPB, XPBC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13872, 2071, 291703 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19447, P49135, Q4G005 | |
Ercc3 | |
ERCC3 Fusion Protein Ag0876 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction