Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC6L Mouse anti-Human, Unconjugated, Clone: 3F12-2B10, Abnova™

Catalog No. 89335034 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-034 Supplier Abnova Corporation Supplier No. H00054821M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ERCC6L Monoclonal antibody specifically detects ERCC6L in Human samples. It is validated for S-ELISA, ELISA, WB-Re, WB-Tr.

Specifications

Antigen ERCC6L
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F12-2B10
Conjugate Unconjugated
Description excision repair cross-complementing rodent repair deficiency, complementation group 6-like
Gene ERCC6L
Gene Accession No. BC008808
Gene Alias FLJ20105, MGC131695, PICH
Host Species Mouse
Immunogen ERCC6L (AAH08808, 1 a.a. ∼ 419 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 54821
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.